Millex® Glass Fiber Syringe Filter
Supplier: Merck
SLAP05010
SLAP05010EA 89.4 EUR
SLAP05010
Millex® Glass Fiber Syringe Filter
Filters Syringe Filters
A 50 mm diameter non-sterile syringe filter with a 2 µm pore size glass fiber membrane.
Order Now
SPECIFICATIONS
Environmentally Preferable
Airflow rate ≥650 L/min air flow rate at 10 psi
4,1 bar max. inlet pressure (60 psi)
Max. temperature 45 °C