Sterilin™ Scintillation Vials with Cap
Supplier: STERILIN
S31
SLINS31EA 595 EUR
SLINS31
Sterilin™ Scintillation Vials with Cap
Vials
Sterilin® Scintillation vials are ideal for use in liquid scintillation counting, beta/gamma counting and for sample storage. Vials are made of PP and caps are PE. The quarter-turn cap ensures reliable sample storage.
  • Precision manufactured for strength and chemical resistance
  • Quarter turn screw cap ensures excellent sample containment
  • 6 ml vial with flush fitting cap is ideal as insert for scintillation vials
  • Not available in all geographic regions
Certifications: Certificate of Analysis (COA), Certificate of Conformance (COC).