DNA marker, pBR322 DNA/BsuRI (HaeIII) marker
Supplier: Thermo Fisher Scientific
SM0271
SM0271EA 67.75 EUR
SM0271
DNA marker, pBR322 DNA/BsuRI (HaeIII) marker
Electrophoresis Reagents Molecular Weight Markers
pBR322 DNA/BsuRI (HaeIII) marker is suitable for sizing and approximate quantification of small linear double-stranded DNA fragments in agarose and non denaturing polyacrylamide gels.
  • Agarose gel compatible
  • Sharp bands
Delivery information: Supplied with loading dye for sample DNA.
Caution: For research use only. Not for use in diagnostic procedures.