Gas diffuser stone
Supplier: SAINT GOBAIN QUARTZ CERAMICS
3590001A
SOBQ3590001AEA 200 EUR
SOBQ3590001A
Gas diffuser stone
Environmental Test Kits
Spherical (25 mm) fine porous aerator stone made of Alundum® fused crystalline aluminium oxide.
Delivery information: Supplied with a cadmium-plated brass nipple fitting serrated for 5 mm rubber tubing.