L-Alanine USP

Supplier: Spectrum Chemical

Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine

A1253-1KG A1253-25KGBL A1253-5KG
SPCMA1253-1KGEA 688 EUR
SPCMA1253-1KG SPCMA1253-25KGBL SPCMA1253-5KG
L-Alanine USP
L-Alanine
Formula: C₃H₇NO₂
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1,4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR