Antimony(III) oxide, Reagent Grade

Supplier: Spectrum Chemical

A1330-12KG A1330-50KGBL
SPCMA1330-12KGEA 1790 EUR
SPCMA1330-12KG SPCMA1330-50KGBL
Antimony(III) oxide, Reagent Grade
Antimony (III) oxide

Antimony Trioxide, Reagent is an inorganic compound and crucial as a commercial compound of antimony. It is an opacifying agent for glasses, ceramics and enamels. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: O₃Sb₂
Storage Temperature: Ambient
MDL Number: MFCD00011214
CAS Number: 1309-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR