Aluminium chloride hexahydrate USP

Supplier: Spectrum Chemical
Warning

Synonyms: Aluminum chloride hexahydrate

AL225-12KG AL225-2.5KG AL225-25KGBL AL225-50KGBL
SPCMAL225-12KGEA 2600 EUR
SPCMAL225-12KG SPCMAL225-2.5KG SPCMAL225-25KGBL SPCMAL225-50KGBL
Aluminium chloride hexahydrate USP
Aluminium chloride hexahydrate
Formula: AlCl₃·6H₂O
Melting Pt: ∼100 °C
Density: 2,39 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149134
CAS Number: 7784-13-6
EINECS: 231-208-1
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 95% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR