Amantadine hydrochloride USP

Supplier: Spectrum Chemical

Synonyms: 1-Adamantylamine hydrochloride, 1-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 1-Aminotricyclo(3.3.1.1(3,7))decan HCl, 1-Aminoadamantane hydrochloride, 1-Adamantanammonium chloride, 1-Adamantanamine hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-1-ylamine hydrochloride, Adamantan-1-amine hydrochloride, 1-Adamantaneammonium Chloride

AM102-1KGBL
SPCMAM102-1KGBLEA 0 EUR
SPCMAM102-1KGBL
Amantadine hydrochloride USP
Amantadine hydrochloride
Formula: C₁₀H₁₈ClN
Melting Pt: 360 °C
MDL Number: MFCD00074723
CAS Number: 665-66-7
EINECS: 211-560-2

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR