2-Hydroxy-4-methoxy-5-sulphobenzophenone ≥97.0%

Supplier: Spectrum Chemical

B1082-12KGBL
SPCMB1082-12KGBLEA 0 EUR
SPCMB1082-12KGBL
2-Hydroxy-4-methoxy-5-sulphobenzophenone ≥97.0%
2-Hydroxy-4-methoxy-5-sulfobenzophenone

Benzophenone-4 is also known as sulisobenzone and is typically an ingredient in most sunscreens. It is used as a skin protectant from damage by short-wave UVA ultraviolet light and UVB.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₄H₁₂O₆S
Storage Temperature: Ambient
MDL Number: MFCD00024962
CAS Number: 4065-45-6

Order Now

Specification Test Results

Assay (Dry Basis) 97.0 - 103.0%
Extinction Coefficient in 95% Ethanol at 285 NM 460 min.
pH of a 1% Solution 1.2 - 2.2
Color (Gardner; 10% W/V in Water) max. 4.0
Solubility of 10% W/V in Water To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR