Cyclopentanone 99

Supplier: Spectrum Chemical

C1124-200LTBL C1124-20LTBL
SPCMC1124-200LTBLEA 23830 EUR
SPCMC1124-200LTBL SPCMC1124-20LTBL
Cyclopentanone 99
Cyclopentanone

Cyclopentanone is a liquid that has an odor similar to peppermint and is devoid of color. It's an organic compound and a cyclic ketone that most often used as a fragrance.


GHS hazard : Flame;Exclamation mark
UN2245

Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 130
Melting Pt: -58
Density: 0,951
Flash Pt: 31
Storage Temperature: Ambient
MDL Number: MFCD00001409
CAS Number: 120-92-3
UN: 2245
ADR: 3 III
Merck Index: 13,02771

Order Now

Specification Test Results

Assay 99 % min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR