Calcium-D-gluconate monohydrate ≥98.5% USP

Supplier: Spectrum Chemical

Synonyms: D-Gluconic acid calcium salt monohydrate, Calcium gluconate monohydrate, Gluconic acid calcium salt monohydrate

CA146-12KG CA146-2.5KG CA146-50KGBL
SPCMCA146-12KGEA 1280 EUR
SPCMCA146-12KG SPCMCA146-2.5KG SPCMCA146-50KGBL
Calcium-D-gluconate monohydrate ≥98.5% USP
Calcium D-gluconate monohydrate
Formula: C₁₂H₂₂CaO₁₄·H₂O
Melting Pt: 178 °C
MDL Number: MFCD00150787
CAS Number: 66905-23-5
EINECS: 206-075-8
Merck Index: 13,01671

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR