Calcium stearate NF

Supplier: Spectrum Chemical

CA169-12KG CA169-2.5KG CA169-45KGBL
SPCMCA169-12KGEA 1150 EUR
SPCMCA169-12KG SPCMCA169-2.5KG SPCMCA169-45KGBL
Calcium stearate NF
Calcium stearate

BSE/TSE statement (available upon request)
Kosher certified

Formula: C₃₆H₇₀CaO₄
Melting Pt: 147…149 °C
Density: 1,065 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036390
CAS Number: 1592-23-0
EINECS: 216-472-8
Merck Index: 13,01708
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66064c431fc65dcdf98ff2bc1d0227f37de78108&vid=c1765b9467ae214d145f3824443319aa43d8bd1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 9% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR