EDTA (ethylenediamine tetraacetic acid) 99,4, powder ACS

Supplier: Spectrum Chemical
Warning

Synonyms: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II

E1040-500GM
SPCME1040-500GMEA 288 EUR
SPCME1040-500GM
EDTA (ethylenediamine tetraacetic acid) 99,4, powder ACS
EDTA (Ethylenediamine tetraacetic acid)

(Ethylenedinitrilo)tetraacetic Acid, Powder, Reagent, ACS, is generally abbreviated as EDTA, a colorless, water soluble aminopolycarboxylic acid and solid. It is used by many industries and in the laboratory for applications ranging from a sequestering agent to chelation therapy. As an ACS grade Reagent, this Spectrum Chemical manufactured compound is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Formula: C₁₀H₁₆N₂O₈
MW: 292.25 g/mol
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 220 °C
Density: 0,86 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.4% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR