Ethyl 4-hydroxybenzoate NF

Supplier: Spectrum Chemical

ET117-12KG ET117-2.5KG ET117-50KGBL
SPCMET117-12KGEA 2510 EUR
SPCMET117-12KG SPCMET117-2.5KG SPCMET117-50KGBL
Ethyl 4-hydroxybenzoate NF
Ethyl 4-hydroxybenzoate
Formula: C₉H₁₀O₃
Boiling Pt: 297 °C (1013 hPa)
Melting Pt: 116…118 °C
Density: 1,078 g/cm³ (131 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002353
CAS Number: 120-47-8
EINECS: 204-399-4
Merck Index: 13,03869

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR