Folic acid ≥97.0%, powder USP

Supplier: Spectrum Chemical

FO105-1KG FO105-25KGBL FO105-5KGBL FO105-100GM
SPCMFO105-1KGEA 4400 EUR
SPCMFO105-1KG SPCMFO105-25KGBL SPCMFO105-5KGBL SPCMFO105-100GM
Folic acid ≥97.0%, powder USP
Folic acid

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₉H₁₉N₇O₆
Storage Temperature: Ambient
MDL Number: MFCD00079305
CAS Number: 59-30-3

Order Now

Specification Test Results

Assay (Anhydrous Basis) 97.0 - 102.0%
Water Determination max.8.5 %
Residue On Ignition max. 0.3 %
Related Compounds max. 2.0 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR