D-(+)-Glucosamine hydrochloride ≥98%

Supplier: Spectrum Chemical

Synonyms: 2-Amino-2-deoxy-D-glucopyranose Hydrochloride, D-Glucosamine hydrochloride, (2R,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride

GL110-1KG GL110-25KGBL GL110-50KGBL GL110-5KG
SPCMGL110-1KGEA 1220 EUR
SPCMGL110-1KG SPCMGL110-25KGBL SPCMGL110-50KGBL SPCMGL110-5KG
D-(+)-Glucosamine hydrochloride ≥98%
D-(+)-Glucosamine hydrochloride

D-Glucosamine Hydrochloride is an amino sugar and plays an important role in the biochemical synthesis of glycosylated lipids and proteins. It is most commonly used as a non-vitamin dietary supplement.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₃NO₅
MW: 215.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00135831
CAS Number: 66-84-2

Order Now

Specification Test Results

Assay 98% min.
Moisture max.1.5 %
Heavy Metals max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR