Hydroxypropyl methylcellulose (Hypromellose), 4000 mPa.s USP Substitution Type 2910

Supplier: Spectrum Chemical

Synonyms: Methocel K100, Hydroxypropyl methyl cellulose

H1335-125GM
SPCMH1335-125GMEA 0 EUR
SPCMH1335-125GM
Hydroxypropyl methylcellulose (Hypromellose), 4000 mPa.s USP Substitution Type 2910
Hydroxypropyl methyl cellulose (Hypromellose)

Hypromellose, Substitution Type 2910, 4000 mPa.s, USP is an artificial tear used to relieve dry eye conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₃H₈O₂·xCH₄O
Storage Temperature: Ambient
MDL Number: MFCD00131360
CAS Number: 9004-65-3

Order Now

Specification Test Results

Viscosity of a 2% Aqueous Solution @ 20D °C 3000- 5600 mPa·s
Assay (Methoxy Groups) 28.0 - 30.0%
Assay (Hydroxypropoxy Groups) 7.0 - 12.0%
pH 5.0 - 8.0
Loss On Drying max. 5.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR