D-Lactose monohydrate, powder NF

Supplier: Spectrum Chemical

LA106-12KG LA106-2.5KG LA106-45KGBL
SPCMLA106-12KGEA 1080 EUR
SPCMLA106-12KG SPCMLA106-2.5KG SPCMLA106-45KGBL
D-Lactose monohydrate, powder NF
D-Lactose monohydrate

Lactose, Monohydrate, Powder, NF is used to help form tablets because it has excellent compressibility properties and as a diluent powder for dry-powder inhalations.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Formula: C₁₂H₂₂O₁₁·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150747
CAS Number: 64044-51-5

Order Now

Specification Test Results

Clarity and Color of Solution To Pass Test
Specific Rotation [A]D +54.4 to +55.9
Acidity / Alkalinity To Pass Test
Water 4.5 - 5.5 %
Loss On Drying max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR