Polyvinylpyrrolidone, M.W. ∼ 40,000 K-30 FCC

Supplier: Spectrum Chemical

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

P1216-500GM
SPCMP1216-500GMEA 483 EUR
SPCMP1216-500GM
Polyvinylpyrrolidone, M.W. ∼ 40,000 K-30 FCC
Polyvinylpyrrolidone

Polyvinylpyrrolidone K-30, FCC is used as an iodine antiseptic. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR