Pyridoxine hydrochloride ≥98% FCC

Supplier: Spectrum Chemical

P1459-100GM P1459-1KG
SPCMP1459-100GMEA 475 EUR
SPCMP1459-100GM SPCMP1459-1KG
Pyridoxine hydrochloride ≥98% FCC
Pyridoxine hydrochloride

Pyridoxine Hydrochloride, FCC, a form of vitamin B6, is used as a vitamin B6 dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₈H₁₂ClNO₃
Melting Pt: 214…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00012807
CAS Number: 58-56-0
EINECS: 200-386-2
Merck Index: 13,08072

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR