Pyridoxine hydrochloride ≥98.0% USP

Supplier: Spectrum Chemical

Synonyms: Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI

PY103-100GM PY103-1KG
SPCMPY103-100GMEA 376 EUR
SPCMPY103-100GM SPCMPY103-1KG
Pyridoxine hydrochloride ≥98.0% USP
Pyridoxine hydrochloride

Pyridoxine Hydrochloride, USP is a form of vitamin B6. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₈H₁₂ClNO₃
MW: 205.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012807
CAS Number: 58-56-0
Merck Index: 13,08072

Order Now

Specification Test Results

Assay (C₈H₁₁NO₃[-HCl) 98.0 - 102.0 %
Chloride Content 16.9 - 17.6 %
Loss On Drying max. 0.5 %
Residue After Ignition max. 0.1 %
Heavy Metals max. 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR