Sodium sulphate, anhydrous ≥99.0% FCC

Supplier: Spectrum Chemical

S1454-500GM S1454-2.5KG S1454-12KG
SPCMS1454-500GMEA 134 EUR
SPCMS1454-500GM SPCMS1454-2.5KG SPCMS1454-12KG
Sodium sulphate, anhydrous ≥99.0% FCC
Sodium sulphate

Sodium Sulfate, Anhydrous, FCC is used as an acidity regulator. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Na₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 100.5%
Lead (Pb) max. 2 mg/kg
Loss On Drying max. 1 %
Selenium (Se) max. 0.003 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR