Sucralose FCC

Supplier: Spectrum Chemical

Synonyms: Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside

S3437-2.5KG S3437-100GM S3437-500GM
SPCMS3437-2.5KGEA 5300 EUR
SPCMS3437-2.5KG SPCMS3437-100GM SPCMS3437-500GM
Sucralose FCC
Sucralose

Sucralose, FCC is used as an artificial sweetener products that require a longer shelf life. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₁₉Cl₃O₈
MW: 397.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03648615
CAS Number: 56038-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR