Triethanolamine (trolamine) NF

Supplier: Spectrum Chemical

TR143-200LTBL TR143-20LT TR143-4LTGL
SPCMTR143-200LTBLEA 7360 EUR
SPCMTR143-200LTBL SPCMTR143-20LT SPCMTR143-4LTGL
Triethanolamine (trolamine) NF
Triethanolamine (Trolamine)

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formula: C₆H₁₅NO₃
Boiling Pt: 335,4 °C (1013 hPa)
Melting Pt: 21,6 °C
Density: 1,13 g/cm³ (20 °C)
Flash Pt: 179 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR