Vitamin A acetate USP

Supplier: Spectrum Chemical

Synonyms: all-trans-Retinyl acetate, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate

V1257-25KGBL
SPCMV1257-25KGBLEA 0 EUR
SPCMV1257-25KGBL
Vitamin A acetate USP
all-trans-Retinyl acetate

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formula: C₂₂H₃₂O₂
MW: 328,49 g/mol
MDL Number: MFCD00019413
CAS Number: 127-47-9
EINECS: 204-844-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR