Hollow fibre filter modules, ME membrane, MidiKros®
Supplier: Spectrum Laboratories
D04-M20U-06-N D04-M20U-10-S D04-M20U-10-N D06-M20U-06-N D06-M20U-10-S D06-M20U-06-S D06-M10U-06-S D06-M10U-06-N D04-M10U-06-S D06-M20U-10-N D04-M20U-06-S D04-M10U-06-N D02-M20U-10-S D02-M20U-10-N D02-M20U-06-S D02-M20U-06-N D02-M10U-06-S D02-M10U-06-N
SPECD04-M20U-06-NEA 767 EUR
SPECD04-M20U-06-N SPECD04-M20U-10-S SPECD04-M20U-10-N SPECD06-M20U-06-N SPECD06-M20U-10-S SPECD06-M20U-06-S SPECD06-M10U-06-S SPECD06-M10U-06-N SPECD04-M10U-06-S SPECD06-M20U-10-N SPECD04-M20U-06-S SPECD04-M10U-06-N SPECD02-M20U-10-S SPECD02-M20U-10-N SPECD02-M20U-06-S SPECD02-M20U-06-N SPECD02-M10U-06-S SPECD02-M10U-06-N
Hollow fibre filter modules, ME membrane, MidiKros®
Filters Tangential Flow Systems
Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applications to full-scale production.
  • Hollow fibre membranes are anisotropic in structure
  • Filter housings are engineered to handle higher pressures and autoclaving
  • Low protein binding increases the product yield of each processing run
  • These filters have FLL/FLL connections and come in three effective lengths
Tangential flow filtration is used in both microfiltration and ultra-filtration applications. Ultra-More Product Information