Hollow fibre filter modules, PES membrane, MidiKros® TC
Supplier: Spectrum Laboratories
T02-P20U-10-N T02-P20U-10-S T04-P20U-10-S T04-P20U-10-N T06-P20U-10-S T06-P20U-10-N
SPECT02-P20U-10-NEA 596 EUR
SPECT02-P20U-10-N SPECT02-P20U-10-S SPECT04-P20U-10-S SPECT04-P20U-10-N SPECT06-P20U-10-S SPECT06-P20U-10-N
Hollow fibre filter modules, PES membrane, MidiKros® TC
Filters Tangential Flow Systems
Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applications to full-scale production.
  • Hollow fibre membranes are anisotropic in structure
  • The filter housings are engineered to handle higher pressures and autoclaving
  • Low protein binding increases the product yield of each processing run
  • TC filters have ¹/₂" TC/FLL connections and come in three effective lengths
Tangential flow filtration is used in both microfiltration and ultra-filtration applications. Ultra-More Product Information