Caspase-3/7 Activity Plate Reader Assay Kit, Green

Supplier: STEMCELL Technologies

100-0920
STMC100-0920EA 861 EUR
STMC100-0920
Caspase-3/7 Activity Plate Reader Assay Kit, Green
Assays Cellular Assays

For detection of caspase-3/7 activity in cell lysates prepared from mammalian cells undergoing apoptosis - 200 tests.


  • Z-DEVD-Rh 110-DVED-Z detects caspase-3/7 activity
  • Excitation wavelength: 500 nm | Emission wavelength: 522 nm
  • Extinction coefficient: 80,000 cm⁻¹M⁻¹
  • Easily scalable for high-throughput assays
  • Verified for use with fluorescent plate readers


Efficiently detect caspase-3/7 activity in cell lysates prepared from mammalian cells undergoing apoptosis by using the Caspase-3/7 Activity Plate Reader Assay Kit, Green, which includes the fluorogenic substrate Z-DEVD-Rh 110-DVED-Z and an assay buffer. This reagent kit uses a quick and simple staining protocol that is well suited for high-throughput screening and compatible with standard fluorescent plate readers.


Caspase-3 and caspase-7 have a proteolytic substrate selectivity for the peptide sequence Asp-Glu-Val-Asp (DEVD). When added to apoptotic cells, Z-DEVD-Rh 110-DVED-Z is cleaved by activated caspase-3/7, generating a green fluorescent signal that can be detected by standard plate readers.


Caspases are a family of proteases that play a central role in cellular apoptosis through the cleavage of select proteins, and result in the disassembly of the cell (Thornberry and Lazebnik Science, 1998). This process is switched on by pro-apoptotic signals from death receptors or stimuli such as cytotoxic reagents, which activate the initiator caspases and lead to activation of the effector caspases-caspase-3 and caspase-7 (Thornberry and Lazebnik Science, 1998). Caspase assays are useful for quantifying caspase-3/7 activity in apoptotic cells and for screening caspase-3/7 inhibitors.


CAS. number: 67-68-5


Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR