EasySep™ RBC Depletion Reagents, 10 ml
Supplier: STEMCELL Technologies
18170 18170RF
STMC18170EA 364 EUR
STMC18170 STMC18170RF
EasySep™ RBC Depletion Reagents, 10 ml
Cell Separation Media
Immunomagnetic isolation of untouched human leukocytes from whole blood.
  • 99,9% RBC depletion without the need for density gradient centrifugation, sedimentation or lysis
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ RBC Depletion Reagent is designed to immunomagnetically deplete red blood cells (RBCs) from fresh human peripheral blood, umbilical cord blood, bone marrow or leukapheresis samples by negative selection. RBCs are targeted with Tetrameric Antibody Complexes recognising Glycophorin A and dextran-coated magneticMore Product Information