EasySep™ Mouse CD4+CD25+ Regulatory T Cell Isolation Kit II
Supplier: STEMCELL Technologies
18783 18783RF
STMC18783EA 1740 EUR
STMC18783 STMC18783RF
EasySep™ Mouse CD4+CD25+ Regulatory T Cell Isolation Kit II
Cell Separation Media
Immunomagnetic isolation of mouse CD4+CD25+ regulatory T cells.
  • Fast and easy-to-use
  • Up to 93% purity
  • No columns required
The EasySep™ Mouse CD4+CD25+ Regulatory T Cell Isolation Kit II is designed to isolate CD4+CD25+ T cells from single-cell suspensions of splenocytes or other tissues by negative followed by positive selection. CD4+ T cells are first enriched by negative selection using the EasySep™ Mouse CD4+ T Cell Isolation Cocktail. CD25+ cells are then positively selected from the pre-enriched cells using EasySep™ Mouse CD25 Regulatory T CellMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable