EasySep™ Human CD8+ T Cell Enrichment Kits
Supplier: STEMCELL Technologies
19053 19053RF
STMC19053EA 1630 EUR
STMC19053 STMC19053RF
EasySep™ Human CD8+ T Cell Enrichment Kits
Cell Separation Media
Immunomagnetic isolation of untouched human CD8+ T cells.
  • Fast, easy-to-use and column-free
  • Up to 95% purity
  • Untouched, viable cells
The EasySep™ Human CD8+ T Cell Enrichment Kit is designed to isolate CD8+ T cells from fresh or previously frozen peripheral blood mononuclear cells or ammonium chloride-lysed leukapheresis sample by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-CD8+ T cells and glycophorin A and dextran-coated magnetic particles. The labelled cells are separated using anMore Product Information