EasySep™ Mouse Monocyte Isolation Kits
Supplier: STEMCELL Technologies
19861 19861RF
STMC19861EA 1560 EUR
STMC19861 STMC19861RF
EasySep™ Mouse Monocyte Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched mouse monocytes
  • Fast and easy-to-use
  • Up to 95% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse Monocyte Isolation Kit targets non-monocyte cells by labeling unwanted cells with antibodies and magnetic particles, and separates cells without columns using an EasySep™ magnet. Desired cells are simply poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, culture or cell-based assays.
This product replaces the EasySep™ MouseMore Product Information