RPMI 1640 Medium
Supplier: STEMCELL Technologies
36750
STMC36750EA 132 EUR
STMC36750
RPMI 1640 Medium
Cell Culture Media
RPMI 1640 medium.
RPMI 1640 Medium is useful for a wide variety of cell culture applications. Selection of a suitable nutrient medium is dependent on cell type, culture conditions, and degree of chemical definition required for the cell culture application.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').