Cole-Parmer® Glass Capillary Tubes
Supplier: Cole Parmer
03013-63 03013-64 03013-65
STUA03013-63EA 36 EUR
STUA03013-63 STUA03013-64 STUA03013-65
Cole-Parmer® Glass Capillary Tubes
Tubes Melting Point Tubes and Capillaries
Accessories for your melting point apparatus.
  • Constructed of soda glass and easily sealed in a Bunsen flame
  • Supplied in a durable tube holder for added protection
Capillary tubes measure 100 mm L, 1,9 mm OD, 1,3 mm ID, and 0,3 mm wall thickness.