Cole-Parmer® Stuart RS-200 Roller Shakers
Supplier: Cole Parmer
51901-24 51901-18 51901-20 51901-22 04653-09
STUA51901-24EA 0 EUR
STUA51901-24 STUA51901-18 STUA51901-20 STUA51901-22 STUA04653-09
Cole-Parmer® Stuart RS-200 Roller Shakers
Shakers Rotating/Rocking Shakers
Gentle rocking and rolling action for complete mixing.
  • Safe for use in cold rooms down to 39 °F (4 °C) or in incubators to 104 °F (40 °C), up to 80% humidity
  • 360° rotation
  • Continuous and quiet operation
  • Side panels ensure sample bottles stay on the shaker
Solidly constructed roller shakers are ideal for mixing blood samples, viscous substances, and liquid-solid suspensions where minimum aeration is required. Select shakers with six or nine tube rollers to mix multiple samples at the same time while saving benchtop space. Order the stacking system 04653More Product Information