Trifluoroacetic acid 99%, Reagent Grade, Fluka™

Supplier: Honeywell Chemicals
Danger

T6508-10AMP T6508-100ML T6508-1L T6508-25ML T6508-500ML T6508-5X10AMP
T-6508EA 100.96 EUR
T-6508 T6508-100ML T6508-1L T6508-25ML T6508-500ML T6508-5X10AMP
Trifluoroacetic acid 99%, Reagent Grade, Fluka™
Trifluoroacetic acid

  • ≥95% for general laboratory use.


Beilstein Number: 742035

Formula: C₂HF₃O₂
Boiling Pt: 72 °C (1,013 hPa)
Melting Pt: -15 °C
Density: 1,48 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Pressure 97.5 mmHg ( 20 °C)
Assay (alkalimetric) 98.5 - 101.5 %
Assay (GC) Min. 98.5 %
Identity (IR) complying
APHA Max. 30
Refractive Index n20/D 1.3 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR