2,3,5-Triphenyltetrazolium chloride (TTC) ≥98% (by HPLC), Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning

Synonyms: Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC

M-Clarity™ quality level = MQ200

T8877-100G T8877-5G T8877-10G T8877-25G T8877-50G
T8877-100GEA 526 EUR
T8877-100G T8877-5G T8877-10G T8877-25G T8877-50G
2,3,5-Triphenyltetrazolium chloride (TTC) ≥98% (by HPLC), Sigma-Aldrich®
2,3,5-Triphenyltetrazolium chloride
Formula: C₁₉H₁₅ClN₄
MW: 334.81 g/mol
Boiling Pt: ∼360 °C (1013 hPa)
Melting Pt: 235
Storage Temperature: Refrigerator
MDL Number: MFCD00011963
CAS Number: 298-96-4
EINECS: 206-071-6
Merck Index: 13,09815
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c333c35973a3c23046a4cf82f13618dcc1c7d9ac&vid=64898dcfe1e696e6f65ede885a0d2331e18e3b01&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR