Cole Parmer® Dri-Block® BH-250 Heaters
Supplier: Cole-Parmer
36620-00 36620-10 36620-04 36620-02 36620-06 36620-08 36620-12 36620-14 36620-16
TECN36620-00EA 983 EUR
TECN36620-00 TECN36620-10 TECN36620-04 TECN36620-02 TECN36620-06 TECN36620-08 TECN36620-12 TECN36620-14 TECN36620-16
Cole Parmer® Dri-Block® BH-250 Heaters
Block Heaters Digital Block Heaters
Choose a heater, then blocks to suit your application needs!
  • Precision-engineered heaters with excellent temperature accuracy
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Take up less storage space-lightweight and flat-top surface allow units to be stacked when not in use
Use these heaters as a dry, constant temperature source for various lab and research applications. All models have independent indicator lights to show main power, heater action, and over-temperature cut out.
The 'number of blocksMore Product Information