Chest Freezers, −25 to −45 °C, LT C
Supplier: TENAK
N111004 N111002 N111006 N111003 N111005
TENAN111004EA 2760 EUR
TENAN111004 TENAN111002 TENAN111006 TENAN111003 TENAN111005
Chest Freezers, −25 to −45 °C, LT C
Freezers Laboratory / General Purpose Freezers
Range of chest freezers with a temperature range −25 to −45 °C.
  • Integrated handle lock
  • Castors
  • Choice of sizes: Personal freezer, small, medium or large chest freezer models
  • Low cost of ownership
  • Small units have compact footprint
  • Medium and large sized units are ideal for use as main storage
These chest freezers have low power consumption but offer excellent stability, reliability, and long-term durability. There is no maintenance required, with exception of defrosting and vacuum cleaning the compressor compartment. Also, no regular service is
More Product Information