Laboratory Chest Freezers, XLT
Supplier: TENAK
N112001 N112002 N122001
TENAN112001EA 3600 EUR
TENAN112001 TENAN112002 TENAN122001
Laboratory Chest Freezers, XLT
Freezers Laboratory / General Purpose Freezers
Laboratory chest freezers with a temperature range −40 to −60 °C. The undercounter model XLT U100 has a temperature range of −25 to −65 °C.
  • Pull-down time less than 100 min for personal freezer XLT C75
  • Low cost of ownership
  • Excellent stability and reliability
  • Long-term durability
  • Castors on XLT C75 and XLT C150 models
  • Undercounter Freezer XLT U100 is stackable
  • XLT C150 is an ideal personal freezer, features an integrated handle lock
  • XLT U100 has 2 port holes and whisper quiet noise level
The XLT C75 and XLT C150 chest freezer models have low power
More Product Information