Vane anemometers, testo 417
Supplier: TESTO
0563.0417 0563.1417 0563.2417
TEST0563.0417EA 385 EUR
TEST0563.0417 TEST0563.1417 TEST0563.2417
Vane anemometers, testo 417
Anemometers
The compact 100 mm vane anemometer testo 417 is the ideal solution for simple, fast and precise determination of flow velocity and volume flow at air intake and outlet vents of air conditioning and ventilation systems.
  • Easy, fast and precise measurement of flow, volume flow and temperature at air inlets and outlets
  • Efficient regulation of balanced residential ventilation and fast documentation with the testo Smart App
  • Fast in-app configuration, graph history, second screen and measurement data memory in the testo Smart App
  • Timed and point mean value
More Product Information