Infrared thermometers, testo 830
Supplier: TESTO
0560.8312 0560.8314 0560.8311 0563.8314
620-2808EA 102 EUR
620-2808 TEST0560.8314 620-2466 TEST0563.8314
Infrared thermometers, testo 830
Thermometers Digital Thermometers
These infrared thermometers are designed for temperature measurements in trade and industry.
  • Non invasive
  • Adjustable alarm limits
  • Fast recording: Two measurements per second
  • Spectral range: 8 - 14 μm
830-T1 infrared thermometer features 1-point laser sighting, 10:1 optics, adjustable limit values and alarm function.
830-T2 infrared thermometer features 2-point laser sighting, 12:1 optics, adjustable limit values, alarm function and connection of external probes.
830-T4 infrared thermometer features 2-point laser sighting, 30:1 optic, adjustableMore Product Information