Multi-position magnetic stirrers, MIXdrive MTP series
Supplier: 2MAG
90250 90252 43006 43012 43024 43096
TMAG90250EA 830 EUR
TMAG90250 TMAG90252 TMAG43006 TMAG43012 TMAG43024 TMAG43096
Multi-position magnetic stirrers, MIXdrive MTP series
Stirrers Magnetic Stirrers, Multi-position
Multi-position magnetic stirrers with hermetically sealed stainless steel housing which is water-, germ- and dust-proof. Suitable for shaking micotitre plates in CO2 incubators and ovens.
  • Maintenance- and wear-free inductive drive
  • 100% synchronised jerk-free mixing
  • Submersible in water and oil baths
MIXcontrol MTP is an external control unit made from easy to clean, resistant stainless steel housing. It includes SoftStart for reliable and safe acceleration of the stirring bar and QuickSet for setting of start and maximum speed. It has automatic memoryMore Product Information