Anion exchange chromatography columns, TSK-GEL®, BioAssist Q
Supplier: TOSOH Bioscience
0019685
TOYO0019685EA 2390 EUR
TOYO0019685
Anion exchange chromatography columns, TSK-GEL®, BioAssist Q
Chromatography Columns
TSK-Gel® BioAssist Q are polymer based packed PEEK columns for laboratory applications. These columns provide high capacity for small to very large MW proteins and nucleic acids.
  • High capacity even for large proteins
  • Unique pore structure