TSKgel® universal reversed phase columns
Supplier: TOSOH Bioscience
0022723 0022713 0022714 0022715 0022716 0022717 0022718 0022719 0022720 0022722 0022710 0022724 0022725 0022726 0022727 0022728 0022729 0022730 0022731 0022721 0022701 0021995 0021996 0022712 0022700 0022711 0022702 0022703 0022704 0022705 0022706 0022707 0022708 0022709 0022734 0021997 0022732 0022757 0022758 0022759 0022760 0022761 0022762 0022763 0022755 0022765 0022754 0022764 0022744 0022735 0022736 0022737 0022738 0022739 0022740 0022741 0022756 0022743 0022733 0022745 0022746 0022747 0022748 0022749 0022750 0022751 0022752 0022753 0022742 0021458 0021459 0021460 0021461 0021462 0021456 0021455 0021810 0021811 0021812 0021813 0021814 0021457 0021829 0021453 0021454 0021938 0021939 0021940 0021830 0021831 0021838 0021839 0021841 0021842 0021843 0021844
TOYO0022723EA 743 EUR
TOYO0022723 TOYO0022713 TOYO0022714 TOYO0022715 TOYO0022716 TOYO0022717 TOYO0022718 TOYO0022719 TOYO0022720 TOYO0022722 TOYO0022710 TOYO0022724 TOYO0022725 TOYO0022726 TOYO0022727 TOYO0022728 TOYO0022729 TOYO0022730 TOYO0022731 TOYO0022721 TOYO0022701 TOYO0021995 TOYO0021996 TOYO0022712 TOYO0022700 TOYO0022711 TOYO0022702 TOYO0022703 TOYO0022704 TOYO0022705 TOYO0022706 TOYO0022707 TOYO0022708 TOYO0022709 TOYO0022734 TOYO0021997 TOYO0022732 TOYO0022757 TOYO0022758 TOYO0022759 TOYO0022760 TOYO0022761 TOYO0022762 TOYO0022763 TOYO0022755 TOYO0022765 TOYO0022754 TOYO0022764 TOYO0022744 TOYO0022735 TOYO0022736 TOYO0022737 TOYO0022738 TOYO0022739 TOYO0022740 TOYO0022741 TOYO0022756 TOYO0022743 TOYO0022733 TOYO0022745 TOYO0022746 TOYO0022747 TOYO0022748 TOYO0022749 TOYO0022750 TOYO0022751 TOYO0022752 TOYO0022753 TOYO0022742 TOYO0021458 TOYO0021459 TOYO0021460 TOYO0021461 TOYO0021462 TOYO0021456 TOYO0021455 TOYO0021810 TOYO0021811 TOYO0021812 TOYO0021813 TOYO0021814 TOYO0021457 TOYO0021829 TOYO0021453 TOYO0021454 TOYO0021938 TOYO0021939 TOYO0021940 TOYO0021830 TOYO0021831 TOYO0021838 TOYO0021839 TOYO0021841 TOYO0021842 TOYO0021843 TOYO0021844
TSKgel® universal reversed phase columns
Chromatography Columns
TSKgel ODS-100V/ODS-100Z columns incorporate the best-inclass surface properties to limit secondary interactions of basic, acidic and chelating compounds.
  • The ultra purity Type B base silica contains negligible amounts of metal ion impurities, minimizes sample adsorption
  • High surface area (450 m²/g) silica
  • Spherical 3 and 5 µm particles with 10 nm (100 Å) pores
  • Very high column efficiency and moderate column back pressure
  • Monomeric bonding chemistry
  • Low residual silanol content
TSKgel ODS-100V provides strong retention for polar compounds due to itsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable