Anion exchange resins, Toyopearl® and TSK-GEL®

Supplier: TOSOH Bioscience

0007472 0007473 0007988 0014026 0014693 0014710 0017223 0017227 0017231 0043201 0043205 0043271 0043281 0043283 0043381 0043383 0014694 0019804 0018535 0018536 0019823 0014692 0014704 0017224 0017228 0017232 0043275 0021855 0021854 0021856 0018546
TOYO0007472EA 1500 EUR
TOYO0007472 TOYO0007473 TOYO0007988 TOYO0014026 TOYO0014693 TOYO0014710 TOYO0017223 TOYO0017227 TOYO0017231 TOYO0043201 TOYO0043205 TOYO0043271 TOYO0043281 TOYO0043283 TOYO0043381 TOYO0043383 TOYO0014694 TOYO0019804 554-5788 TOYO0018536 TOYO0019823 TOYO0014692 TOYO0014704 TOYO0017224 TOYO0017228 TOYO0017232 TOYO0043275 TOYO0021855 TOYO0021854 TOYO0021856 TOYO0018546
Anion exchange resins, Toyopearl® and TSK-GEL®
Chromatography Sorbents HPLC Sorbents

Anion exchange resins are polymer-based media developed for the separation of biomolecules including monoclonal antibodies and oligonucleotides. Several of these resins are available in multiple particle sizes allowing for their use in capture, intermediate purification or polishing process steps.


TOYOPEARL DEAE-650 is a weak anion exchange resin for biomolecule purification. It has high chemical and mechanical stability, allowing cleaning-in-place (CIP) and very high flow rates. It is frequently used in plasma fractionation and for the purification of recombinant blood coagulation factors.
TOYOPEARL SuperQ-650 media is a strong anion exchanger for biomolecule purification. It can be used for high throughput capture, intermediate purification, and polishing process steps.


TOYOPEARL GigaCap Q-650 media are high capacity anion exchange resins optimised for the capture and purification of both small and large proteins, including oligonucleotides and monoclonal antibodies. This media is specifically designed for packed bed use.
TOYOPEARL GigaCap DEAE-650M is a very high capacity anion exchange resin.


TOYOPEARL QAE-550 resin is a strong anion exchanger useful for purification of proteins up to 100 kD. It is used for high throughput capture of feedstocks.
TOYOPEARL NH2-750F media is a salt tolerant anion exchange resin capable of aggregate removal in both flow-through and binding and elution modes. This resin is ideal for process scale applications from the capture of proteins from biological feedstock.


TSKgel SuperQ-5PW media is a strong anion exchanger whereas TSKgel DEAE-5PW media is a weak anion for biomolecule purification. Its small particle size makes it an excellent choice for polishing and high-performance resolution steps.

Order Now

SPECIFICATIONS

Type DEAE-650C DEAE-650M SuperQ-650M DEAE-5PW (30) SuperQ-5PW (30) DEAE-5PW (20) SuperQ-5PW (20) DEAE-650S SuperQ-650S QAE-550C SuperQ-650C GigaCap Q-650M
Environmentally Preferable
Amount of ionic groups 50 - 110 µeq/ml 80 - 120 µeq/ml 200 - 300 µeq/ml 50 - 110 µeq/ml 120 - 180 µeq/ml 50 - 110 µeq/ml 120 - 180 µeq/ml 80 - 120 µeq/ml 200 - 300 µeq/ml 280 - 380 µeq/ml 200 - 300 µeq/ml 200 - 300 µeq/ml
Average particle size 50 - 150 µm 40 - 90 µm 40 - 90 µm 20 - 40 µm 20 - 40 µm 15 - 25 µm 15 - 25 µm 20 - 50 µm 20 - 50 µm 50 - 150 µm 50 - 150 µm 50 - 100 µm
Pressure resistance 3 bar 3 bar 3 bar 3 bar 3 bar 3 bar 3 bar 3 bar 3 bar 3 bar 3 bar 3 bar
Working pH 2 - 13 2 - 13 2 - 13 2 - 13 2 - 13 2 - 13 2 - 13 2 - 13 2 - 13 2 - 13 2 - 13 2 - 13

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR