Dithiothreitol (DTT, Cleland's reagent) 0.1 mol/l in aqueous solution

Supplier: TREVIGEN

4145-100-02
TREV4145-100-02EA 0 EUR
TREV4145-100-02
Dithiothreitol (DTT, Cleland's reagent) 0.1 mol/l in aqueous solution
Dithiothreitol (DTT, Cleland's reagent)
Formula: C₄H₁₀O₂S₂
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR