Two-piece fingertight fittings for ¹/₁₆" Ø tubing, SealTight™, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
F-192 F-192X F-193 F-195 F-195X F-196 F-350
UPCHF-192EA 12.4 EUR
UPCHF-192 UPCHF-192X UPCHF-193 UPCHF-195 UPCHF-195X UPCHF-196 UPCHF-350
Two-piece fingertight fittings for ¹/₁₆" Ø tubing, SealTight™, Upchurch Scientific®
Fittings and Connectors
Several nut lengths and head styles to fit into a variety of applications, designed to connect ¹/₁₆" OD tubing to 10-32 coned ports, hold up to 9000 psi (620 bar).
The dual compression created by the specially designed nut and ferrule enables the SealTight fittings system to outperform standard finger tightened fittings. The forward cone of
the SealTight Ferrule provides gripping power and a leak-free seal via conventional compression by the receiving port. The slotted end creates the second compression zone in conjunction with a SealTight nut, to be used with the F-192 ferrule.