One-piece fittings, Super Flangeless™, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
P-249
UPCHP-249EA 25.5 EUR
UPCHP-249
One-piece fittings, Super Flangeless™, Upchurch Scientific®
Fittings and Connectors
One piece super flangeless fitting is ideal for those connections that need to be broken frequently and tubing assemblies because they swage onto the tubing.
  • Excellent chemical resistance