Extender tools
Supplier: UPCHURCH SCIENTIFIC
P-399 N-290 P-277 P-278 P-291 P-292 P-297 P-298 P-299
UPCHP-399EA 39.8 EUR
UPCHP-399 UPCHN-290 UPCHP-277 UPCHP-278 UPCHP-291 UPCHP-292 UPCHP-297 UPCHP-298 UPCHP-299
Extender tools
Wrenches
These tools made from aluminium or Delrin® can be used to tighten most knurled nuts in hard to reach places. For precise tightening, the extender tools listed with ¼" hex drives are designed to adapt to any torque wrench with a female ¼" socket.